4-Chloro-α,α-dimethylphenylacetic acid 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(4-Chlorophenyl)-2-methylpropanoic acid

L05277.03 L05277.06
L05277.03EA 15.1 GBP
L05277.03 L05277.06
4-Chloro-α,α-dimethylphenylacetic acid 98%
4-Chloro-alpha,alpha-dimethylphenylacetic acid

Purity: 98% CAS No.: 6258-30-6 Molecular Formula: C10H11ClO2 Formula Weight: 198.65 Melting Point: 124-129° Beilstein Reference: 2094056 MDL No.: MFCD00044814

Formula: C₁₀H₁₁ClO₂
Melting Pt: 124…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00044814
CAS Number: 6258-30-6
EINECS: 228-392-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR