Ethyl hydrogen suberate 98%

Supplier: Thermo Scientific
Warning

Synonyms: 8-Ethoxy-8-oxooctanoic acid

L04272.06 L04272.14
L04272.06EA 77.8 GBP
L04272.06 L04272.14
Ethyl hydrogen suberate 98%
Ethyl hydrogen suberate

Purity: 98% CAS No.: 14113-01-0 Molecular Formula: CH3CH2O2C(CH2)6CO2H Formula Weight: 202.25 Melting Point: 25-27° Beilstein Reference: 1775737 MDL No.: MFCD00014385

Formula: CH₃CH₂O₂C(CH₂)₆CO₂H
MW: 202,25 g/mol
Melting Pt: 25…27 °C
Storage Temperature: Ambient
MDL Number: MFCD00014385
CAS Number: 14113-01-0
EINECS: 237-968-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR