Cefmetazole
Supplier: Thermo Fisher Scientific
|
Danger
|
A broad spectrum second generation cephalosporin antibiotic.
CAS No.: 56796-20-4
Molecular Formula: C15H17N7O5S3
Formula Weight: 471.53
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water
MDL No.: MFCD00865068
Applications: A broad spectrum second generation cephalosporin antibiotic
Formula:
C₁₅H₁₇N₇O₅S₃ |
MDL Number:
MFCD00865068 CAS Number: 56796-20-4 EINECS: 260-384-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...