Cefamandole nafate
Supplier: Thermo Fisher Scientific
|
Danger
|
J66735.06
J66735.09
J66735.06EA
0
GBP
J66735.06
J66735.09
Cefamandole nafate
Cefamandole nafate
CAS No.: 42540-40-9
Molecular Formula: C19H18N6NaO6S2
Formula Weight: 513.50
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 190°
Solubility: Freely soluble in water. Sparingly soluble in methanol
MDL No.: MFCD00864877
Applications: A second generation cephalosporin antibiotic.
Formula:
C₁₉H₁₇N₆NaO₆S₂ MW: 512.491 g/mol |
MDL Number:
MFCD00864877 CAS Number: 42540-40-9 EINECS: 255-877-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...