Minocycline hydrochloride 890-950 µg/mg

Supplier: Thermo Scientific
Warning

Synonyms: (4S,4aS,5aR,12aS)-4,7-Bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride, Minocycline HCl

J66429.ME J66429.03
J66429.MEEA 226 GBP
J66429.ME J66429.03
Minocycline hydrochloride 890-950 µg/mg
Minocycline hydrochloride

CAS No.: 13614-98-7 Molecular Formula: C23H27N3O7.HCl Formula Weight: 493.94 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: Yellow Melting Point: 205-210° Boiling Point: 813° Solubility: Soluble in water MDL No.: MFCD00083669


Applications: A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections

Formula: C₂₃H₂₇N₃O₇
MW: 493,94 g/mol
Boiling Pt: 813 °C
Melting Pt: 205…210 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00083669
CAS Number: 13614-98-7
EINECS: 237-099-7
Merck Index: 14,6202

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR