Dexamethasone sodium phosphate 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Dexamethasone 21-Phosphate disodium salt hydrate, Dexamethasone 21-phosphate disodium salt

J64083.03 J64083.06
J64083.03EA 97.1 GBP
J64083.03 J64083.06
Dexamethasone sodium phosphate 98%
Dexamethasone sodium phosphate

Purity: 98% CAS No.: 2392-39-4 Molecular Formula: C22H28FNa2O8P Formula Weight: 516.40 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Melting Point: 233-240° Beilstein Reference: 6473066 MDL No.: MFCD00079105

Formula: C₂₂H₂₈FNa₂O₈P
MW: 516.41 g/mol
Melting Pt: 233…240 °C
MDL Number: MFCD00079105
CAS Number: 2392-39-4
EINECS: 219-243-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR