Temsirolimus ≥99%
Supplier: Thermo Scientific
Synonyms:
(1R,2R,4R)-4-((R)-2-((3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,27-Dihydroxy-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-1,5,11,28,29-pentaoxo-1,4,5,6,9,10,11,12,13,14,21,22,23,24,25,26,27,28,29,31,32,33,34,34a-tetracosahydro-3H-23,27-epoxypyrido[2,1-c][1]oxa[4]azacyclohentriacontin-3-yl)propyl)-2-methoxycyclohexyl 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoate
Purity: 99+%
CAS No.: 162635-04-3
Molecular Formula: C56H87NO16
Formula Weight: 1030.29
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00934421
Applications: A Rapamycin derivative with anti-cancer activity that inhibits mTOR
Formula:
C₅₆H₈₇NO₁₆ MW: 1030,29 g/mol |
MDL Number:
MFCD00934421 CAS Number: 162635-04-3 Merck Index: 14,9142 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...