Aspergillus niger Pectinase
Supplier: Thermo Scientific
J63408.MD
J63408.03
J63408.MDEA
121
GBP
J63408.MD
J63408.03
Aspergillus niger Pectinase
Enzymes
CAS No.: 9032-75-1
Physical Form: Powder
Appearance: Off-white to pale brown
MDL No.: MFCD00131809
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...