Naproxen sodium 98%
Supplier: Thermo Fisher Scientific
Synonyms:
Sodium (S)-2-(6-methoxynaphthalen-2-yl)propanoate
J63103.06
J63103.14
J63103.22
J63103.06EA
77.5
GBP
J63103.06
J63103.14
J63103.22
Naproxen sodium 98%
Naproxen sodium
Purity: 98%
CAS No.: 26159-34-2
Molecular Formula: C14H13NaO3
Formula Weight: 252.24
Physical Form: Powder
Appearance: White
Melting Point: 259-262°
Solubility: Soluble in water up to 100mg/ml
MDL No.: MFCD00058507
Applications: A cyclooxygenase inhibitor
Formula:
C₁₄H₁₃NaO₃ MW: 252.24 g/mol Melting Pt: 259…262 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058507 CAS Number: 26159-34-2 EINECS: 247-486-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...