Camostat methanesulphate 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Camostat Mesylate, Camostat Methanesulphonate, 4-(2-(2-(Dimethylamino)-2-oxoethoxy)-2-oxoethyl)phenyl 4-guanidinobenzoate methanesulphonate
J62362.MC
J62362.ME
J62362.MCEA
265
GBP
J62362.MC
J62362.ME
Camostat methanesulphate 98%
Camostat methanesulphate
Purity: 98%
CAS No.: 59721-29-8
Molecular Formula: C20H22N4O5.CH3SO3H
Formula Weight: 494.52
Physical Form: Powder
Appearance: White
Melting Point: 164-168°
MDL No.: MFCD00941410
Applications: An orally active synthetic serine protease inhibitor
Formula:
C₂₀H₂₂N₄O₅·CH₃SO₃H MW: 494,52 g/mol Melting Pt: 164…168 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00941410 CAS Number: 59721-29-8 UN: 3077 ADR: 9,III Merck Index: 14,1728 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...