Bumetanide ≥98%
Supplier: Thermo Scientific
J62302.03
J62302.06
J62302.03EA
116
GBP
J62302.03
J62302.06
Bumetanide ≥98%
Bumetanide
Purity: 98+%
CAS No.: 28395-03-1
Molecular Formula: C17H20N2O5S
Formula Weight: 364.40
Physical Form: Solid
Appearance: White
Melting Point: 230-231°
MDL No.: MFCD00078949
Applications: Inhibitor of the sodium-potassium-chloride cotransporter
Formula:
C₁₇H₂₀N₂O₅S Melting Pt: 230…231 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078949 CAS Number: 28395-03-1 EINECS: 249-004-6 Merck Index: 14,1484 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...