5-Iodouridine 96%
Supplier: Thermo Fisher Scientific
Synonyms:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione
J62259.03
J62259.06
J62259.03EA
76.1
GBP
J62259.03
J62259.06
5-Iodouridine 96%
5-Iodouridine
Purity: 96%
CAS No.: 1024-99-3
Molecular Formula: C9H11IN2O6
Formula Weight: 370.10
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 33665
MDL No.: MFCD00006532
Formula:
C₉H₁₁IN₂O₆ |
MDL Number:
MFCD00006532 CAS Number: 1024-99-3 EINECS: 213-833-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...