5-Hydroxydecanoic acid sodium salt 98%
Supplier: Thermo Scientific
J61434.MC
J61434.ME
J61434.MCEA
119
GBP
J61434.MC
J61434.ME
5-Hydroxydecanoic acid sodium salt 98%
5-Hydroxydecanoic acid sodium salt
Purity: 98%
CAS No.: 624-00-0
Molecular Formula: C10H19NaO3
Formula Weight: 210.25
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00153808
Applications: A potassium channel antagonist which blocks the postischemic effects of the potassium channel activator cromakalim
Formula:
C₁₀H₁₉NaO₃ MW: 210,25 g/mol |
MDL Number:
MFCD00153808 CAS Number: 624-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...