Jenner's stain stain for hematology
Supplier: Thermo Fisher Scientific
Synonyms:
Eosin−Methylene Blue
J61077.14
J61077.22
J61077.14EA
20.5
GBP
J61077.14
J61077.22
Jenner's stain stain for hematology
Jenner's stain
CAS No.: 62851-42-7
Physical Form: Powder
Appearance: Dark green
MDL No.: MFCD00081733
Applications: For use in staining blood smears
Storage Temperature:
Ambient |
MDL Number:
MFCD00081733 CAS Number: 62851-42-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...