Iota carrageenan

Supplier: Thermo Scientific

J60603.22 J60603.30
J60603.22EA 53.6 GBP
J60603.22 J60603.30
Iota carrageenan
Iota carrageenan

CAS No.: 9062-07-1 Physical Form: Powder Appearance: White to pale brown MDL No.: MFCD00151512


Applications: Forms soft gels in the presence of calcium ions

Storage Temperature: Ambient
MDL Number: MFCD00151512
CAS Number: 9062-07-1
EINECS: 232-949-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR