Canrenone 98%
Supplier: Thermo Fisher Scientific
J60238.03
J60238.06
J60238.14
J60238.03EA
54.9
GBP
J60238.03
J60238.06
J60238.14
Canrenone 98%
Canrenone
Purity: 98%
CAS No.: 976-71-6
Molecular Formula: C22H28O3
Formula Weight: 340.46
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 149-151°
MDL No.: MFCD00071735
Applications: Inhibits aldosterone biosynthesis and blocks ouabain effects
Formula:
C₂₂H₂₈O₃ Melting Pt: 149…151 °C |
MDL Number:
MFCD00071735 CAS Number: 976-71-6 EINECS: 213-554-5 Merck Index: 14,1752 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...