Caronic anhydride 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3,3-Dimethyl-1,2-cyclopropanedicarboxylic Anhydride, 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione
H66892.14
H66892.22
H66892.14EA
65.3
GBP
H66892.14
H66892.22
Caronic anhydride 98%
Caronic anhydride
Purity: 98%
CAS No.: 67911-21-1
Molecular Formula: C7H8O3
Formula Weight: 140.14
Physical Form: Solid
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 53-57°
Boiling Point: 137°/14mm
Beilstein Reference: 117391
MDL No.: MFCD09751198
Formula:
C₇H₈O₃ MW: 140,14 g/mol Boiling Pt: 137 °C (14 mmHg) Melting Pt: 53…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD09751198 CAS Number: 67911-21-1 EINECS: 614-164-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...