3,4-Methylenedioxyphenethylamine 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-[3,4-(Methylenedioxy)phenyl]ethylamine
H66445.03
H66445.06
H66445.03EA
45
GBP
H66445.03
H66445.06
3,4-Methylenedioxyphenethylamine 95%
3,4-Methylenedioxyphenethylamine
Purity: 95%
CAS No.: 1484-85-1
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Beilstein Reference: 383838
MDL No.: MFCD00060509
Formula:
C9H11NO2 MW: 165.19 g/mol |
MDL Number:
MFCD00060509 CAS Number: 1484-85-1 EINECS: 216-060-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...