L-Cyclopropylglycine 97%
Supplier: Thermo Scientific
Synonyms:
2-Cyclopropyl-L-glycine, (S)-2-Amino-2-cyclopropylacetic acid, H-Cyclopropyl-Gly-OH
H65755.MD
H65755.03
H65755.06
H65755.MDEA
43.3
GBP
H65755.MD
H65755.03
H65755.06
L-Cyclopropylglycine 97%
L-Cyclopropylglycine
Purity: 97%
CAS No.: 49606-99-7
Molecular Formula: C5H9NO2
Formula Weight: 115.13
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
Appearance: White to off-white
Beilstein Reference: 5728866
Optical Rotation: +101°(c=1 in 1M HCl)
MDL No.: MFCD06659116
Formula:
C₅H₉NO₂ MW: 115,132 g/mol |
MDL Number:
MFCD06659116 CAS Number: 49606-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...