[4-(4-Pyridyl)thiazole-2-yl]acetonitrile 97%
Supplier: Thermo Scientific
|
Warning
|
H65261.MD
H65261.03
H65261.MDEA
44.8
GBP
H65261.MD
H65261.03
[4-(4-Pyridyl)thiazole-2-yl]acetonitrile 97%
[4-(4-Pyridyl)thiazole-2-yl]acetonitrile
Purity: 97%
CAS No.: 1016888-78-0
Molecular Formula: C10H7N3S
Formula Weight: 201.25
Physical Form: Solid
Appearance: Yellow to brown
UN No.: UN3439
MDL No.: MFCD09950233
Formula:
C₁₀H₇N₃S Storage Temperature: Ambient |
MDL Number:
MFCD09950233 CAS Number: 1016888-78-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...