4-Phenyl-2-thiazoleacetonitrile 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-(4-Phenylthiazol-2-yl)acetonitrile
H65040.MD
H65040.03
H65040.MDEA
0
GBP
H65040.MD
H65040.03
4-Phenyl-2-thiazoleacetonitrile 97%
4-Phenyl-2-thiazoleacetonitrile
Purity: 97%
CAS No.: 41381-89-9
Molecular Formula: C11H8N2S
Formula Weight: 200.26
Physical Form: Solid
Appearance: Pale brown
UN No.: UN3439
MDL No.: MFCD00551791
Formula:
C₁₁H₈N₂S Storage Temperature: Ambient |
MDL Number:
MFCD00551791 CAS Number: 41381-89-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...