(3-(Pyridin-3-yl)phenyl)boronic acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-(3-Pyridyl)benzeneboronic acid, [3-(Pyridin-3-yl)phenyl]boronic acid
H64739.MD
H64739.MDEA
18.2
GBP
H64739.MD
(3-(Pyridin-3-yl)phenyl)boronic acid 98%
(3-(Pyridin-3-yl)phenyl)boronic acid
Purity: 98%
CAS No.: 351422-72-5
Molecular Formula: C11H10BNO2
Formula Weight: 199.02
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 20520996
MDL No.: MFCD16658918
Formula:
C₁₁H₁₀BNO₂ MW: 199,016 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16658918 CAS Number: 351422-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...