4(1H)-Pyrimidinone 98%
Supplier: Thermo Scientific
Synonyms:
3,4-Dihydropyrimidin-4-one, 1H-Pyrimidin-4-one, 4-Hydroxypyrimidine, Pyrimidin-4(1H)-one
H64690.03
H64690.06
H64690.03EA
24.3
GBP
H64690.03
H64690.06
4(1H)-Pyrimidinone 98%
4(1H)-Pyrimidinone
Purity: 98%
CAS No.: 4562-27-0
Molecular Formula: C4H4N2O
Formula Weight: 96.09
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 163-169° lit.
Beilstein Reference: 741972
MDL No.: MFCD00006664
Formula:
C₄H₄N₂O MW: 96,09 g/mol Melting Pt: 163…169 °C (lit.) |
MDL Number:
MFCD00006664 CAS Number: 51953-17-4 EINECS: 224-932-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...