5-Bromopyrimidine-2-carboxylic acid 97%

Supplier: Thermo Fisher Scientific
Warning

H64041.03 H64041.06
H64041.03EA 0 GBP
H64041.03 H64041.06
5-Bromopyrimidine-2-carboxylic acid 97%
5-Bromopyrimidine-2-carboxylic acid

Purity: 98% CAS No.: 37131-87-6 Molecular Formula: C5H3BrN2O2 Formula Weight: 203.00 Physical Form: Solid Melting Point: 193-198° Beilstein Reference: 121432 MDL No.: MFCD00496793

Formula: C₅H₃BrN₂O₂
Melting Pt: 193…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00496793
CAS Number: 37131-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR