(3-(Naphthalen-1-yl)phenyl)boronic acid 98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-(1-Naphthyl)phenylboronic acid, 3-(1-Naphthyl)benzeneboronic acid

H64019.MD H64019.03 H64019.06
H64019.MDEA 20.2 GBP
H64019.MD H64019.03 H64019.06
(3-(Naphthalen-1-yl)phenyl)boronic acid 98%
(3-(Naphthalen-1-yl)phenyl)boronic acid

Purity: 98% CAS No.: 881913-20-8 Molecular Formula: C16H13BO2 Formula Weight: 248.09 Physical Form: Powder Appearance: Off-white Beilstein Reference: 18694809 MDL No.: MFCD11045054

Formula: C₁₆H₁₃BO₂
MW: 248,088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11045054
CAS Number: 881913-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR