2,3-Dimethylphenylacetic acid 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,3-Dimethylbenzeneacetic acid
H63947.03
H63947.06
H63947.03EA
49.6
GBP
H63947.03
H63947.06
2,3-Dimethylphenylacetic acid 95%
2,3-Dimethylphenylacetic acid
Purity: 95%
CAS No.: 30981-98-7
Molecular Formula: C10H12O2
Formula Weight: 164.2
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD03844736
Formula:
C₁₀H₁₂O₂ MW: 164,204 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03844736 CAS Number: 30981-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...