D-4,4'-Biphenylalanine 98%

Supplier: Thermo Fisher Scientific

Synonyms: 4-Phenyl-D-phenyalanine, H-4-Phenyl-D-Phe-OH, 3-(4-Biphenylyl)-D-alanine, (R)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid

H63785.MD H63785.03
H63785.MDEA 0 GBP
H63785.MD H63785.03
D-4,4'-Biphenylalanine 98%
D-4,4'-Biphenylalanine

Purity: 98% CAS No.: 170080-13-4 Molecular Formula: C15H15NO2 Formula Weight: 241.29 Physical Form: Powder Appearance: White MDL No.: MFCD01318879

Formula: C₁₅H₁₅NO₂
MW: 241.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318879
CAS Number: 170080-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR