Methyl-6-bromo-1-methyl-1H-indole-2-carboxylate 97%

Supplier: Thermo Scientific

Synonyms: Methyl 6-bromo-1-methylindole-2-carboxylate

H63655.06
H63655.06EA 343 GBP
H63655.06
Methyl-6-bromo-1-methyl-1H-indole-2-carboxylate 97%
Methyl-6-bromo-1-methyl-1H-indole-2-carboxylate

Purity: 97% CAS No.: 680569-18-0 Molecular Formula: C11H10BrNO2 Formula Weight: 268.11 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Pale Yellow MDL No.: MFCD07364734

Formula: C₁₁H₁₀BrNO₂
MW: 268,11 g/mol
MDL Number: MFCD07364734
CAS Number: 680569-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR