D-Tyrosine methyl ester hydrochloride 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
H-D-Tyr-OMe.HCl, H-D-Tyr-OMe·HCl
H63630.06
H63630.14
H63630.06EA
52.6
GBP
H63630.06
H63630.14
D-Tyrosine methyl ester hydrochloride 98%
D-Tyrosine methyl ester hydrochloride
Purity: 98%
CAS No.: 3728-20-9
Molecular Formula: C10H13NO3.HCl
Formula Weight: 231.68
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00066124
Formula:
C₁₀H₁₄ClNO₃ MW: 231,676 g/mol |
MDL Number:
MFCD00066124 CAS Number: 3728-20-9 EINECS: 223-084-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...