(2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(benzyloxy)pyrrolidine-2-carboxylic acid 98%

Supplier: Thermo Scientific

Synonyms: trans-4-Benzyloxy-N-Fmoc-L-proline, Fmoc-Hyp(Bzl)-OH

H63582.MD H63582.03
H63582.MDEA 29 GBP
H63582.MD H63582.03
(2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(benzyloxy)pyrrolidine-2-carboxylic acid 98%
(2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(benzyloxy)pyrrolidine-2-carboxylic acid

Purity: 98% CAS No.: 174800-02-3 Molecular Formula: C27H25NO5 Formula Weight: 443.49 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD00237023

Formula: C₂₇H₂₅NO₅
MW: 443,499 g/mol
MDL Number: MFCD00237023
CAS Number: 174800-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR