Boc-D-phenylalanine methyl ester 98%

Supplier: Thermo Scientific

Synonyms: (R)-Methyl 2-((tert-butoxycarbonyl)amino)-3-phenylpropanoate, Boc-D-Phe-OMe, N-Boc-D-phenylalanine methyl ester

H63466.03 H63466.06 H63466.14
H63466.03EA 29.2 GBP
H63466.03 H63466.06 H63466.14
Boc-D-phenylalanine methyl ester 98%
Boc-D-phenylalanine methyl ester

Purity: 98% CAS No.: 77119-84-7 Molecular Formula: C15H21NO4 Formula Weight: 279.34 Physical Form: Solid MDL No.: MFCD00672509

Formula: C₁₅H₂₁NO₄
MW: 279,336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672509
CAS Number: 77119-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR