6-methoxyquinoxaline 98%

Supplier: Thermo Fisher Scientific
Warning

H63396.MD H63396.03 H63396.06
H63396.MDEA 0 GBP
H63396.MD H63396.03 H63396.06
6-methoxyquinoxaline 98%
6-methoxyquinoxaline

Purity: 98% CAS No.: 6639-82-3 Molecular Formula: C9H8N2O Formula Weight: 160.17 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD00666601

Formula: C₉H₈N₂O
MDL Number: MFCD00666601
CAS Number: 6639-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR