2-(Trifluoromethyl)isonicotinic acid 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(Trifluoromethyl)pyridine-4-carboxylic acid
H63315.MD
H63315.03
H63315.MDEA
70.9
GBP
H63315.MD
H63315.03
2-(Trifluoromethyl)isonicotinic acid 97%
2-(Trifluoromethyl)isonicotinic acid
Purity: 97%
CAS No.: 131747-41-6
Molecular Formula: C7H4F3NO2
Formula Weight: 191.11
Physical Form: Solid
Appearance: Off-white
Melting Point: 217-223°
MDL No.: MFCD07774132
Formula:
C₇H₄F₃NO₂ MW: 191,11 g/mol Melting Pt: 217…223 °C Storage Temperature: Ambient |
MDL Number:
MFCD07774132 CAS Number: 131747-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...