2-Bromobenzothiazole 99%

Supplier: Thermo Scientific
Warning

H63286.03 H63286.06 H63286.14
H63286.03EA 44.2 GBP
H63286.03 H63286.06 H63286.14
2-Bromobenzothiazole 99%
2-Bromobenzothiazole

Purity: 99% CAS No.: 2516-40-7 Molecular Formula: C7H4BrNS Formula Weight: 214.08 Flash Point: 110°(230°F) Physical Form: Crystalline solid Appearance: Pale yellow Melting Point: 39-43° MDL No.: MFCD02681887

Formula: C₇H₄BrNS
MW: 214,09 g/mol
Melting Pt: 39…43 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02681887
CAS Number: 2516-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR