Methyl-4-fluoro-2-methylbenzoate 98%

Supplier: Thermo Scientific

Synonyms: Methyl 4-fluoro-2-methylbenzoate

H63268.06 H63268.14 H63268.22
H63268.06EA 41 GBP
H63268.06 H63268.14 H63268.22
Methyl-4-fluoro-2-methylbenzoate 98%
Methyl 4-fluoro-2-methylbenzoate

Purity: 98% CAS No.: 174403-69-1 Molecular Formula: C9H9FO2 Formula Weight: 168.16 Physical Form: Liquid Appearance: Colourless MDL No.: MFCD06203790

Formula: C₉H₉FO₂
MW: 168,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203790
CAS Number: 174403-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR