Boc-D-Glu(OtBu)-OH 98%

Supplier: Thermo Fisher Scientific

Synonyms: N-Boc-D-glutamic acid 5-tert-butyl ester

H63172.03 H63172.06
H63172.03EA 0 GBP
H63172.03 H63172.06
Boc-D-Glu(OtBu)-OH 98%
Boc-D-Glu(OtBu)-OH

Purity: 98% CAS No.: 104719-63-3 Molecular Formula: C14H25NO6 Formula Weight: 303.35 Physical Form: Solid Beilstein Reference: 3594191 MDL No.: MFCD00076927

Formula: C₁₄H₂₅NO₆
MW: 303.355 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076927
CAS Number: 104719-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR