2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-[Bis(3,5-dimethylphenyl)phosphanyl]benzaldehyde, 0, 2-(Bis(3,5-dimethylphenyl)phosphino)benzaldehyde
H63117.MC
H63117.ME
H63117.MCEA
39.4
GBP
H63117.MC
H63117.ME
2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde 97%
2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde
Purity: 97%
CAS No.: 669091-00-3
Molecular Formula: C23H23OP
Formula Weight: 346.40
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: Yellow
Melting Point: 106-110°
MDL No.: MFCD15143662
Formula:
C₂₃H₂₃OP MW: 346,41 g/mol Melting Pt: 106…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD15143662 CAS Number: 669091-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...