trans-1-(Boc-amino)-3-cyanocyclohexane 97%

Supplier: Thermo Scientific
Warning

H62850.MD H62850.03
H62850.MDEA 60 GBP
H62850.MD H62850.03
trans-1-(Boc-amino)-3-cyanocyclohexane 97%
trans-1-(Boc-amino)-3-cyanocyclohexane

Purity: 97% CAS No.: 920966-19-4 Molecular Formula: C12H20N2O2 Formula Weight: 224.29 UN No.: UN3439 MDL No.: MFCD22192325

Formula: C₁₂H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22192325
CAS Number: 920966-19-4
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR