4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester 96%
Supplier: Thermo Fisher Scientific
H62733.MD
H62733.03
H62733.MDEA
0
GBP
H62733.MD
H62733.03
4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester 96%
4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester
Purity: 96%
CAS No.: 1154740-65-4
Molecular Formula: C12H16BClFNO2
Formula Weight: 271.52
Physical Form: Solid
Appearance: White
MDL No.: MFCD16996254
Formula:
C₁₂H₁₆BClFNO₂ MW: 271.523 g/mol |
MDL Number:
MFCD16996254 CAS Number: 1154740-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...