2-(4-Chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: Thermo Scientific

Synonyms: 4-Chloro-3-fluorophenylboronic acid pinacol ester, 4-Chloro-3-fluorobenzeneboronic acid pinacol ester

H62445.MD H62445.03
H62445.MDEA 74 GBP
H62445.MD H62445.03
2-(4-Chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(4-Chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Purity: 96% CAS No.: 627525-83-1 Molecular Formula: C12H15BClFO2 Formula Weight: 256.51 Physical Form: Solid MDL No.: MFCD16996258

Formula: C₁₂H₁₅BClFO₂
MW: 256,508 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996258
CAS Number: 627525-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR