3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific

Synonyms: 5-Fluoro-3-hydroxyphenylboronic acid pinacol ester

H62234.MD H62234.03
H62234.MDEA 145 GBP
H62234.MD H62234.03
3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester 96%
3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C12H16BFO3 Formula Weight: 238.06 Physical Form: Solid

Formula: C₁₂H₁₆BFO₃
MW: 238,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16994409
CAS Number: 1887126-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR