Ethyl-3-oxo-3-(pyridin-4-yl)propanoate 98%

Supplier: Thermo Scientific

Synonyms: Ethyl isonicotinoylacetateEthyl 3-oxo-3-(4-pyridyl)propionateEthyl 3-oxo-3-(pyridin-4-yl)propanoate

H61647.06EA 70.7 GBP
H61647.06
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate 98%
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate
Formula: C₁₀H₁₁NO₃
MW: 193,20 g/mol
Melting Pt: 53…59 °C
Density: 1,155
Storage Temperature: Ambient
MDL Number: MFCD00094021
CAS Number: 26377-17-3
EINECS: 247-635-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR