(R,R)-(-)-2-Amino-1-phenylpropyldiphenylphosphine ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(1R,2R)-1-(Diphenylphosphino)-1-phenylpropan-2-amine
H60639.MD
H60639.03
H60639.MDEA
121
GBP
H60639.MD
H60639.03
(R,R)-(-)-2-Amino-1-phenylpropyldiphenylphosphine ≥97%
(R,R)-(-)-2-Amino-1-phenylpropyldiphenylphosphine
Purity: 97+%
CAS No.: 799297-44-2
Molecular Formula: C21H22NP
Formula Weight: 319.38
Melting Point: 105-109°
Optical Rotation: -140° (c=0.5 in chloroform)
UN No.: UN3259
MDL No.: MFCD17018768
Formula:
C₂₁H₂₂NP MW: 319,388 g/mol Melting Pt: 105…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD17018768 CAS Number: 799297-44-2 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...