(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane 97+%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(+)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ethane, (R,R)-Et-BPE, (+)-1,2-Bis((2R,5R)-2,5-diethylphospholano)ethane
H60447.MD
H60447.03
H60447.MDEA
113
GBP
H60447.MD
H60447.03
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane 97+%
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane
Purity: 97+%
CAS No.: 136705-62-9
Molecular Formula: C18H36P2
Formula Weight: 314.43
Flash Point: 110°(230°F)
Refractive Index: 1.5250
Boiling Point: 104-106°/0.05mm
Optical Rotation: +320° (c=1 in hexane)
UN No.: UN2845
MDL No.: MFCD01073768
Formula:
C₁₈H₃₆P₂ MW: 314,434 g/mol Boiling Pt: 104…106 °C (0,05 mmHg) Density: 0,939 Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01073768 CAS Number: 136705-62-9 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...