(S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine 96%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (S)-1-Cbz-3-aminopyrrolidine, benzyl (3S)-3-aminopyrrolidine-1-carboxylate

H58138.MD H58138.03
H58138.MDEA 0 GBP
H58138.MD H58138.03
(S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine 96%
(S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine

Purity: 96% CAS No.: 122536-72-5 Molecular Formula: C12H16N2O2 Formula Weight: 220.27 Flash Point: 110°(230°F) Refractive Index: 1.5400 Melting Point: 310-316° Optical Rotation: +5° (c=1 in methanol) MDL No.: MFCD06858462

Formula: C₁₂H₁₆N₂O₂
Melting Pt: 310…316 °C
Density: 1,155
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD06858462
CAS Number: 122536-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR