3-Hydroxy-2-pyrrolidinone 95%
Supplier: Thermo Scientific
|
Warning
|
H57961.MD
H57961.03
H57961.MDEA
35.6
GBP
H57961.MD
H57961.03
3-Hydroxy-2-pyrrolidinone 95%
3-Hydroxy-2-pyrrolidinone
Purity: 95%
CAS No.: 15116-68-4
Molecular Formula: C4H7NO2
Formula Weight: 101.11
Storage Temperature: 2°C to 8°C
Melting Point: 102-107°
MDL No.: MFCD09751256
Formula:
C₄H₇NO₂ MW: 101,11 g/mol Melting Pt: 102…107 °C |
MDL Number:
MFCD09751256 CAS Number: 15166-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...