(S)-1-Boc-pyrrolidine-3-carboxylic acid 95%
Supplier: Thermo Scientific
Synonyms:
(S)-1-Boc-Pyrrolidine-3-carboxylic acid, (3S)-1-tert-butoxycarbonylpyrrolidine-3-carboxylic acid
H57888.MD
H57888.03
H57888.MDEA
33.3
GBP
H57888.MD
H57888.03
(S)-1-Boc-pyrrolidine-3-carboxylic acid 95%
(S)-1-Boc-pyrrolidine-3-carboxylic acid
Purity: 95%
CAS No.: 140148-70-5
Molecular Formula: C10H17NO4
Formula Weight: 215.25
Storage Temperature: 2°C to 8°C
Optical Rotation: +15° (c=0.8 in chloroform)
MDL No.: MFCD03094728
Formula:
C₁₀H₁₇NO₄ MW: 215,25 g/mol |
MDL Number:
MFCD03094728 CAS Number: 140148-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...