1-Boc-3-(n-propylamino)azetidine 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
H57725.MD
H57725.03
H57725.MDEA
0
GBP
H57725.MD
H57725.03
1-Boc-3-(n-propylamino)azetidine 95%
1-Boc-3-(n-propylamino)azetidine
Purity: 95%
Molecular Formula: C11H22N2O2
Formula Weight: 214.31
MDL No.: MFCD18326837
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...