2-Chloro-N-ethylnicotinamide 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H57514.MD
H57514.03
H57514.MDEA
0
GBP
H57514.MD
H57514.03
2-Chloro-N-ethylnicotinamide 97%
2-Chloro-N-ethylnicotinamide
Purity: 97%
CAS No.: 52943-22-3
Molecular Formula: C8H9ClN2O
Formula Weight: 184.63
MDL No.: MFCD01358076
Formula:
C₈H₉ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD01358076 CAS Number: 52943-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...