1-Benzylazetidin-3-ol 95%
Supplier: Thermo Scientific
|
Warning
|
H57498.03
H57498.06
H57498.03EA
72.6
GBP
H57498.03
H57498.06
1-Benzylazetidin-3-ol 95%
1-Benzylazetidin-3-ol
Purity: 95%
CAS No.: 54881-13-9
Molecular Formula: C10H13NO
Formula Weight: 163.22
MDL No.: MFCD00963607
Formula:
C₁₀H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD00963607 CAS Number: 54881-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...